Learn More
CENPC Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 14250-1-AP-20UL
 View more versions of this product
Product Code. 16812814
  • 1081.84 DKK / 20µL
This item is not returnable. View return policy
Description
Description
Centromere protein C 1 is a centromere autoantigen and a component of the inner kinetochore plate. The protein is required for maintaining proper kinetochore size and a timely transition to anaphase. A putative pseudogene exists on chromosome 12.
Specifications
Specifications
CENPC
Polyclonal
Unconjugated
CENPC
CENP C, CENP C 1, CENPC, CENPC1, Centromere autoantigen C, centromere protein C 1, hcp 4, ICEN7, MIF2
Rabbit
Antigen Affinity Chromatography
RUO
1060, 12617
-20°C
Liquid
Immunofluorescence, Immunocytochemistry, Western Blot
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q03188
CENPC, Cenpc1
CENPC Fusion Protein Ag5523
20 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CENPC Rabbit anti-Human, Mouse, Polyclonal, Proteintech > 20 μL; Unconjugated