Learn More
WTW™ Chemical Oxygen Demand (COD) Test Kits
Get accurate Chemical Oxygen Demand (COD) test results with the Chlorine dioxide reagent test kit.
Brand:  WTW™ 251992
716.25 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
 View more versions of this product
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery. Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery. Regulatory: This product fall under regulation “Lov om kemiske stoffer og produkter - lovbekendtgørelse nr. 1755 af 22. december 2006. If applicable, customers must provide a license before delivery.
Product Code. 11767105
  • 738.00 DKK / 400mL
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Variety of instrumentation for measuring Chemical Oxygen Demand (COD)
  • Safe determinations of COD in water
  • Chlorine dioxide (ClO<sub>2</sub>) reagent test kit
  • For waste water, production control, ground water, surface water
  • For PhotoLab&reg; S6, PhotoLab&reg; S12, PhotoLab&reg; Spectral, spectroFlex photometers
TRUSTED_SUSTAINABILITY
Specifications
Specifications
EN ISO 15705
PhotoLab™ 6000 photometer, Photoflex™ meter
200 to 15,000 mg/L
Wastewater, production control, ground water, surface water
60
Boron (B) reagent test kit
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
WTW™ Chemical Oxygen Demand (COD) Test Kits > Cuvette Test; Detection Range: 200 to 15000mg/L