missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ CHKtide
A peptide substrate based upon CDC25C that is phosphorylated by CHEK1 and other kinases.
Brand: Thermo Scientific™ PV5368
This item is not returnable.
View return policy
Description
A peptide substrate based upon CDC25C that is phosphorylated by CHEK1 and other kinases.
For use with the Adapta™ Universal Kinase Assay (Cat. No. PV5099).

Specifications
KKKVSRSGLYRSPSMPENLNRPR | |
CHKtide peptide substrate in in 50mM HEPES pH 7.5, 0.01% Brij-35, 10mM MgCl2 and 1mM EGTA. | |
2mM CHKtide peptide substrate in 50 mM HEPES pH 7.5, 0.01% Brij-35, 10 mM MgCl2, and 1 mM EGTA Store at -20°C. |
|
1mL, 1 mL | |
Store at -20°C. | |
RUO - research use only | |
Kinase Substrate | |
KKKVSRSGLYRSPSMPENLNRPR |
Kinase Assay, Industrial and Applied Science, Kinase Assay, Kinase Biology, Pharma and Biopharma, Target and Lead Identification and Validation | |
2701.2 Da | |
Fluorescence | |
Frozen | |
peptide based upon CDC25C | |
Peptide-Based Substrate | |
Serine/Threonine Kinases | |
Dry Ice |
Product Suggestions
Customers who viewed this item also viewed
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Thermo Scientific™ CHKtide > 1 mL
For Research Use Only. Not for use in diagnostic procedures.
Spot an opportunity for improvement?Share a Content Correction