missing translation for 'onlineSavingsMsg'
Learn More
Chloroacetamidine hydrochloride, 96%, Thermo Scientific Chemicals Product Code.: 11402607

Chloroacetamidine hydrochloride, 96%, Thermo Scientific Chemicals

Product Code. 11402607
25 g
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy

Product Code. 11402607

Brand: Thermo Scientific Alfa Aesar A14835.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chloroacetamidine hydrochloride is used as a general pharmacophore in delineating characteristics of the amidinotransferase superfamily.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Chloroacetamidine hydrochloride is used as a general pharmacophore in delineating characteristics of the amidinotransferase superfamily.

Solubility
Insoluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10300-69-3
Molecular Formula C2H6Cl2N2
Molecular Weight (g/mol) 128.98
MDL Number MFCD00053013
InChI Key GUPOZVHRTJYZCX-UHFFFAOYSA-N
Synonym chloroacetamidine hydrochloride, 2-chloroacetimidamide hydrochloride, 2-chloroethanimidamide hydrochloride, 2-chloroacetamidine hydrochloride, 2-chloroacetamidine hcl, 2-chloroacetamidine monohydrochloride, 2-chloroacetamidinehydrochloride, ethanimidamide, 2-chloro-, monohydrochloride, chloracetamidine hydrochloride, chloroacetamidinehclShow MoreShow Less
PubChem CID 2776957
IUPAC Name 2-chloroethanimidamide;hydrochloride
SMILES [H+].[Cl-].NC(=N)CCl

Specifications

Melting Point 97°C to 103°C
Quantity 25 g
UN Number UN3261
Beilstein 3678354
Sensitivity Hygroscopic
Solubility Information Insoluble in water.
Formula Weight 129
Percent Purity 96%
Chemical Name or Material Chloroacetamidine hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10162156 Certificate TypeCertificate of Analysis Date06/07/2022 Catalog NumberA14835.06,A14835.14
Lot NumberUSLF000863 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14835.06,A14835.14
Lot Number10154561 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14835.06,A14835.14
Lot Number10154557 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14835.06,A14835.14
Lot Number10098512 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA14835.06,A14835.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Chloroacetamidine hydrochloride, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.