Learn More
Macherey-Nagel™ Chromabond™ NAN Polypropylene SPE Column
2825.00 DKK - 3570.00 DKK
Product Code | Brand | Column Capacity (Metric) | Bed Weight | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Column Capacity (Metric) | Bed Weight | Price | Quantity & Availability | |||||
10513792
|
Macherey-Nagel™
730109 |
3 mL | 400/1400/400 mg |
3570.00 DKK
Pack of 50 |
Estimated Shipment: 24-07-2025
Log in to see stock. |
|||||
10159770
|
Macherey-Nagel™
730149 |
6 mL | 700/2000/700 mg |
2825.00 DKK
Pack of 30 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Specifications
SPE | |
Solid Phase Extraction Column |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Macherey-Nagel™ Chromabond™ NAN Polypropylene SPE Column