Learn More
Macherey-Nagel™ Chromafix™ HR-XA Cartridge
Brand:  Macherey-Nagel™ 731770
 View more versions of this product
Product Code. 12814511
  • 2475.00 DKK / Pack of 50
Estimated Shipment: 31-01-2025
to see stock.
Explore more special offers
Specifications
Spherical PS/DVB Copolymer
45 or 85 μm
Chromafix HR-XA Cartridge
50 Pack
500 mg
Strong quat. ammonium modified polymeric anion exchanger
55 to 65 Å
850 m2/g