missing translation for 'onlineSavingsMsg'
Learn More

(-)-Cinchonidine, 99% (total base), may cont. up to 5% quinine

Product Code. p-7024337
25 g, 25g
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy
This item is not returnable. View return policy

CAS: 485-71-2 | C19H22N2O | 294.40 g/mol

It finds its uses as an antimalarial compound. It is a lysosomotropic agent, inhibits Plasmodium falciparum, interferes with ferriprotoporphyrin formation and is a weak SERT inhibitor.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It finds its uses as an antimalarial compound. It is a lysosomotropic agent, inhibits Plasmodium falciparum, interferes with ferriprotoporphyrin formation and is a weak SERT inhibitor.

Solubility
Insoluble in water. Soluble in alcohol and chloroform.

Notes
Light Sensitive. Store away from oxidizing agents and light. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 485-71-2
Molecular Formula C19H22N2O
Molecular Weight (g/mol) 294.40
MDL Number MFCD00006783
InChI Key KMPWYEUPVWOPIM-UHFFFAOYNA-N
Synonym cinchonidine
PubChem CID 45358337
SMILES OC(C1CC2CCN1CC2C=C)C1=C2C=CC=CC2=NC=C1

Specifications

Melting Point 201°C to 206°C
Quantity 25 g
Beilstein 89690
Sensitivity Light sensitive
Merck Index 14,2286
Solubility Information Insoluble in water. Soluble in alcohol and chloroform.
Optical Rotation −109° (c=0.5 in Ethanol)
IUPAC Name {5-ethenyl-1-azabicyclo[2.2.2]octan-2-yl}(quinolin-4-yl)methanol
Formula Weight 294.39
Percent Purity 99%
Assay (total base), may cont. up to 5% quinine
Chemical Name or Material (-)-Cinchonidine
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(-)-Cinchonidine, 99% (total base), may cont. up to 5% quinine >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.