missing translation for 'onlineSavingsMsg'
Learn More
cis-1,2,3,6-Tetrahydrophthalimide, 96%, Thermo Scientific™ Product Code.: 10091590

cis-1,2,3,6-Tetrahydrophthalimide, 96%, Thermo Scientific™

Product Code. 10091590 Shop All Thermo Scientific Chemicals Products
250 g, Plastic bottle, 250g
Click to view available options
Quantity:
5 g
100 g
250 g
1 kg
Packaging:
Glass bottle
Plastic bottle
Unit Size:
100g
1kg
250g
5g
This item is not returnable. View return policy

Product Code. 10091590

Brand: Thermo Scientific Acros 138232500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 1469-48-3
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.16
MDL Number MFCD00005880
InChI Key CIFFBTOJCKSRJY-OLQVQODUSA-N
Synonym cis-1,2,3,6-tetrahydrophthalimide, cis-3a,4,7,7a-tetrahydro-1h-isoindole-1,3 2h-dione, cis-4-cyclohexene-1,2-dicarboximide, cis-tetrahydrophthalimide, unii-es57d28o1u, 4-cyclohexene-1,2-dicarboximide, cis, cis-.delta.4-tetrahydrophthalimide, 1h-isoindole-1,3 2h-dione, 3a,4,7,7a-tetrahydro-, cis, 1h-isoindole-1,3 2h-dione, 3a,4,7,7a-tetrahydro-, 3ar,7as-rel, 3ar,7as-3a,4,7,7a-tetrahydroisoindole-1,3-dioneShow MoreShow Less
PubChem CID 92888
IUPAC Name (3aR,7aS)-3a,4,7,7a-tetrahydroisoindole-1,3-dione
SMILES C1C=CCC2C1C(=O)NC2=O

Specifications

Melting Point 131.0°C to 138.0°C
Color Beige to Brown-Yellow or Yellow
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Plastic bottle
Quantity 250 g
Formula Weight 151.16
Percent Purity 96%
Physical Form Powder or Flakes
Chemical Name or Material cis-1,2,3,6-Tetrahydrophthalimide, 96%
compliance-icons
Product Identifier
  • cis-1,2,3,6-Tetrahydrophthalimide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H332-Harmful if inhaled.
  • H312-Harmful in contact with skin.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
cis-1,2,3,6-Tetrahydrophthalimide, 96%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.