missing translation for 'onlineSavingsMsg'
Learn More
cis-4-Hexen-1-ol, 97% Product Code.: 10343250

cis-4-Hexen-1-ol, 97%

Product Code. p-4258992 Shop All Thermo Scientific Chemicals Products
5 mL, Glass bottle, 5mL
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Unit Size:
25mL
5mL
This item is not returnable. View return policy

Product Code. 10343250

Brand: Thermo Scientific Acros 439740050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 928-91-6 | C6H12O | 100.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 928-91-6
Molecular Formula C6H12O
Molecular Weight (g/mol) 100.16
MDL Number MFCD00148974,MFCD00009713
InChI Key VTIODUHBZHNXFP-IHWYPQMZSA-N
Synonym cis-4-hexen-1-ol, z-hex-4-en-1-ol, z-4-hexen-1-ol, 4z-hex-4-en-1-ol, 4-hexen-1-ol, z, unii-780wma6j84, e-4-hexenol, cis-1-hydroxy-4-hexene, cis-4-hexene-1-ol, 4z-4-hexen-1-ol #
PubChem CID 5365589
SMILES C\C=C/CCCO

Specifications

Color Colorless
Density 0.8570g/mL
Boiling Point 70°C (28.0 mmHg)
Flash Point 65°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 mL
Specific Gravity 0.857
IUPAC Name (4Z)-hex-4-en-1-ol
Formula Weight 100.16
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material cis-4-Hexen-1-ol
Show More Show Less
Product Identifier
  • cis-4-Hexen-1-ol, 97%

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
cis-4-Hexen-1-ol, 97% >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.