Learn More
Alfa Aesar™ cis-9-Tricosene, 96%
Catalog Number 11302167
Click to view available options
Quantity:
1 g
5 g
27519-02-4
C23H46
322.621
MFCD00008988
IGOWHGRNPLFNDJ-ZPHPHTNESA-N
cis-9-tricosene, muscalure, z-9-tricosene, z-tricos-9-ene, 9-tricosene, z, muscamone, cis-tricos-9-ene, caswell no. 883c, 9z-tricosene, unii-6bsp6hfw73
5365075
(Z)-tricos-9-ene
CCCCCCCCCCCCCC=CCCCCCCCC
27519-02-4
C23H46
322.621
MFCD00008988
IGOWHGRNPLFNDJ-ZPHPHTNESA-N
cis-9-tricosene, muscalure, z-9-tricosene, z-tricos-9-ene, 9-tricosene, z, muscamone, cis-tricos-9-ene, caswell no. 883c, 9z-tricosene, unii-6bsp6hfw73
5365075
(Z)-tricos-9-ene
CCCCCCCCCCCCCC=CCCCCCCCC
Applications
cis-9-Tricosene acts as an insect attractant (sex pheromone) of the female house fly. Further, it is used for biochemical research. In addition, it is used in fly paper and other traps to lure male flies.
Solubility
Miscible with water and hexane. Immiscible with alcohol.
Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 0.806
Boiling Point 299°C to 300°C
Flash Point >110°C (230°F)
Refractive Index 1.453
Quantity 1 g
Beilstein 1841622
Merck Index 14,6309
Solubility Information Miscible with water and hexane. Immiscible with alcohol.
Formula Weight 322.62
Percent Purity 96%
Chemical Name or Material cis-9-Tricosene