missing translation for 'onlineSavingsMsg'
Learn More
compliance-icons

Citric Acid Anhydrous, ACS Reagent Grade, Scharlab

Product Code. 11339933
1 kg
Click to view available options
Quantity:
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Used as an acidifying agent, in analytical chemistry, as a laboratory reagent, in the food industry and as an antioxidant.

  • Synonyms: 2-Hydroxy-1,2,3-propanetricarboxylic acid, b-Hydroxy tricarboxilic acid
  • Molecular Formula: C6H8O7
  • Molar Mass: 192.13g/mol
  • CAS: 77-92-9
  • EIN ECS No.: 201-069-1

Specifications

CAS 77-92-9
Melting Point ∼153°C (decomposition)
Molecular Formula C6H8O7
MDL Number MFCD00011669
Synonym citric acid, citric acid, anhydrous, citro, anhydrous citric acid, citrate, aciletten, citretten, chemfill, hydrocerol a, 1,2,3-propanetricarboxylic acid, 2-hydroxy
Solubility Information Soluble in water (20°C)
InChI Key KRKNYBCHXYNGOX-UHFFFAOYSA-N
SMILES OC(=O)CC(O)(CC(O)=O)C(O)=O
IUPAC Name 2-hydroxypropane-1,2,3-tricarboxylic acid
Molecular Weight (g/mol) 192.12
PubChem CID 311
ChEBI CHEBI:30769
Formula Weight 192.13g/mol
Percent Purity 99.5% (min.) (acidimetric)
Grade ACS Reagent
Quantity 1 kg
Ignition Point 345°C
Residue after Ignition 0.02% max
Vapor Pressure <0.1 hPa (at 20°C)
Show More Show Less
compliance-icons
Product Identifier
  • Citric acid anhydrous, reagent grade, ACS, Reag. Ph Eur
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P264-Wash thoroughly after handling.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Citric Acid Anhydrous, ACS Reagent Grade, Scharlab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.