Learn More
Hirschmann™ Class A Measuring Cylinders, Blue Graduation, with Plastic Socket
Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann Instruments™ Class A Measuring Cylinders, Blue Graduation, with Plastic Socket are manufactured from durable DURAN™ glass tubing.
166.00 DKK - 610.00 DKK
Specifications
Base Type | Hexagonal |
---|---|
Certifications/Compliance | DIN EN ISO 4788 |
Class | A |
Type | Graduated Measuring Cylinder |
Scale Color | Blue |
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | ||||||
11756692
|
Hirschmann™
2270160 |
10 mL |
166.00 DKK
Pack of 2 |
In Stock
Log in to see stock. |
||||||
11766692
|
Hirschmann™
2270170 |
25 mL |
170.00 DKK
Pack of 2 |
In Stock
Log in to see stock. |
||||||
11776692
|
Hirschmann™
2270175 |
50 mL |
201.00 DKK
Pack of 2 |
In Stock
Log in to see stock. |
||||||
11756702
|
Hirschmann™
2270180 |
100 mL |
218.00 DKK
Pack of 2 |
Estimated Shipment: 28-07-2025
Log in to see stock. |
||||||
11766702
|
Hirschmann™
2270186 |
250 mL |
381.00 DKK
Pack of 2 |
In Stock
Log in to see stock. |
||||||
11776702
|
Hirschmann™
2270190 |
500 mL |
610.00 DKK
Pack of 2 |
In Stock
Log in to see stock. |
||||||
10557961
|
Hirschmann™
2270193 |
1000 mL |
477.00 DKK
Each |
In Stock
Log in to see stock. |
||||||
Description
- With spout and plastic socket, main point ring graduation
- Blue graduation
- Conformity certified, with dated batch information
- Acc. to DIN EN ISO 4788
- Corrosion-free and resistant to thermal shock
Specifications
Hexagonal | |
A | |
Blue | |
Glass |
DIN EN ISO 4788 | |
Graduated Measuring Cylinder | |
Round |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Hirschmann™ Class A Measuring Cylinders, Blue Graduation, with Plastic Socket