Learn More
Hirschmann™ Class A Measuring Cylinders, Schellbach Graduation
Excellent for precise liquid measurement and fluid transfer in the laboratory. Hirschmann Instruments™ Class A Measuring Cylinders, Schellbach Graduation are manufactured from durable DURAN™ glass tubing.
239.00 DKK - 1225.00 DKK
Specifications
Certifications/Compliance DIN EN ISO 4788
Class A
View More Specs
Products 9
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10238283
missing translation for 'viewDocuments'
Hirschmann™
2220160
10 mL
239.00 DKK
Pack of 2
Only null left
In Stock
Add to basket
13157939
missing translation for 'viewDocuments'
Hirschmann™
2220154
5 mL
248.00 DKK
Pack of 2
Only null left
In Stock
Add to basket
11857523
missing translation for 'viewDocuments'
Hirschmann™
2220170
25 mL
244.00 DKK
Pack of 2
Only null left
Estimated Shipment: 16-07-2025
Add to basket
12678665
missing translation for 'viewDocuments'
Hirschmann™
2220186
250 mL
528.00 DKK
Pack of 2
Only null left
In Stock
Add to basket
11886983
missing translation for 'viewDocuments'
Hirschmann™
2220175
50 mL
261.00 DKK
Pack of 2
Only null left
In Stock
Add to basket
12668665
missing translation for 'viewDocuments'
Hirschmann™
2220180
100 mL
323.00 DKK
Pack of 2
Only null left
Estimated Shipment: 16-07-2025
Add to basket
11806993
missing translation for 'viewDocuments'
Hirschmann™
2220190
500 mL
887.00 DKK
Pack of 2
Only null left
Estimated Shipment: 16-07-2025
Add to basket
10687321
missing translation for 'viewDocuments'
Hirschmann™
2220193
1000 mL
590.00 DKK
Each
Only null left
In Stock
Edge
Add to basket
10325611
missing translation for 'viewDocuments'
Hirschmann™
2220194
2000 mL
1225.00 DKK
Each
Only null left
In Stock
Add to basket
Description
Description
  • With spout, main point ring graduation
  • Blue, Schellbach stain graduation
  • Conformity certified, with dated batch certificate
  • Acc. to DIN EN ISO 4788
  • Corrosion-free and resistant to thermal shock
Specifications
Specifications
DIN EN ISO 4788
A
Product Title
Hirschmann™ Class A Measuring Cylinders, Schellbach Graduation