Learn More
Zwiebel™ Class F1 Box Weights
Use with weighing instruments of medium accuracy class II. Zwiebel™ Class F1 Box Weights are ideal for the verification or calibration of class F2 weights.
Brand:  Zwiebel™ PBI23083
 View more versions of this product
Product Code. 12031643
  • 1620.00 DKK / Each
Only null left
Estimated Shipment: 08-08-2025
Description
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Specifications
1000 g
±5.00 mg
For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Zwiebel™ Class F1 Box Weights > Mass: 1000g; Tolerance: ±5.00mg