Learn More
Zwiebel™ Class F2 Box Weights
Brand:  Zwiebel™ PBI5682F
 View more versions of this product
Product Code. 11800922
  • 889.00 DKK / Each
Only null left
Estimated Shipment: 07-08-2025
Specifications
1000 g
±15 mg
For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Zwiebel™ Class F2 Box Weights > Mass: 1000g