Learn More
Zwiebel™ Class F2 Box Weights
Brand:  Zwiebel™ PBI56829
 View more versions of this product
Product Code. 11840912
  • 253.00 DKK / Each
Only null left
Estimated Shipment: 04-08-2025
Specifications
10 g
±0.6 mg
For use with Weighing Instruments of Special Accuracy Class I and High Accuracy Class II
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Zwiebel™ Class F2 Box Weights > Mass: 10g