Learn More
Zwiebel™ Class M1 Box Weights with Certificate
Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M1 Box Weights are ideal for the verification or calibration of class M₂ weights.
Brand:  Zwiebel™ PBI23117
 View more versions of this product
Product Code. 11740763
  • 200.00 DKK / Each
Only null left
Estimated Shipment: 27-08-2025
Description
  • Class M1 stainless steel calibration weight
  • Delivered with a calibration certificate guaranteeing traceability
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Specifications
Specifications
±1.2 mg
For use with Weighing Instruments of Medium Accuracy Class III and Ordinary Accuracy Class IIII
Certificates
View accessory products
Promotions
Promotions
Product Content Correction
Product Title
Zwiebel™ Class M1 Box Weights with Certificate > Mass: 2g