Learn More
Zwiebel™ Class M2 Chrome Weight Set
Use with weighing instruments of medium accuracy class III. Zwiebel™ Class M2 Weights are ideal for the verification or calibration of class M3 weights.
1100.00 DKK - 2355.00 DKK
Products 2
Product Code Brand Mass Price Quantity & Availability  
Product Code Brand Mass Price Quantity & Availability  
11724714
View Documents
Zwiebel™
PBI23705
1 to 1000 g
2355.00 DKK
Each
Only null left
Estimated Shipment: 27-08-2025
Add to basket
11704714
View Documents
Zwiebel™
PBI23703
1 to 200 g
1100.00 DKK
Each
Only null left
Estimated Shipment: 27-08-2025
Add to basket
Description
Description
  • Class M2 stainless steel calibration weight
  • In compliance with OIML R111
  • Supplied individually or as a set, in a plastic case or a wooden box
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Zwiebel™ Class M2 Chrome Weight Set