Learn More
Claudin 1 Rabbit anti-Canine, Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 13050-1-AP-150UL
 View more versions of this product
Product Code. 16811074
  • 3058.99 DKK / 150µL
This item is not returnable. View return policy
Description
Description
Claudin-1 is a member of the transmembrane protein family claudins located in cell-cell tight junctions and it acts as a co-receptor for HCV entry into hepatic cells. Claudins are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Claudin-1 is expressed in most tissues such as bladder, fallopian tube, liver, pancreas, prostate, and skin.
Specifications
Specifications
Claudin 1
Polyclonal
Unconjugated
CLDN1
claudin 1, CLD1, CLDN1, ILVASC, SEMP1
Rabbit
Antigen Affinity Chromatography
RUO
12737, 608207, 9076
-20°C
Liquid
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O88551, O95832
CLDN1
Claudin 1 Fusion Protein Ag3713
150 μL
Primary
Canine, Human, Mouse
Antibody
IgG
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Claudin 1 Rabbit anti-Canine, Human, Mouse, Polyclonal, Proteintech > 150 μL; Unconjugated