Learn More
Sampling Systems™ Cleaning Coupons for Plastics
Test and validate your cleaning regime with this range of cleaning validation coupons for various plastics.
Brand:  Sampling Systems™ 8644N-100
 View more versions of this product
Product Code. 15819551
  • 486.00 DKK / Each
Estimated Shipment: 08-07-2025
Explore more special offers
Description
  • Tests available for HDPE, nylon, PP, PTFE and PVC
  • Two sizes available for each configuration, either 50 x 50mm or 100 x 100mm
Specifications
Specifications
Cleaning Coupon
100 x 100 mm
Testing and validating cleaning regime
Nylon
Documents
One moment while we fetch your results.
Product Title
Sampling Systems™ Cleaning Coupons for Plastics > Material: Nylon; Size: 100 x 100mm