Learn More
CleanSpace™ HALO Half Mask for PAPR Assembly
Specifically designed to work to complete the CleanSpace HALO system and clip quickly onto the power units. Half masks are made of soft silicone that readily molds to the face and does not cause allergic dermatitis.
Brand: CleanSpace™ CS3004
Description
- Used with the revolutionary CleanSpace HALO: light weight, no hoses or belts
- Half masks available in 3 sizes to facilitate a good seal and a comfortable fit
- Materials: Soft silicone cushion, nylon clips and polycarbonate (exhalation valve)
- Mask Fit Test adaptor available for TSI Portacount™ fit testing
- Easy removal
- Compatible with most cleaning and disinfection protocols
- Transparent and facilitates communication
Specifications
AS/NZS1716: 2012 | |
PAPR Half Mask | |
Half mask only | |
No |
EN 12942 | |
Medium | |
Primary healthcare facilities, pharmaceutical production, laboratories, animal research facilities and emergency/disaster responders | |
CS3004 |
If you require intrinsically safe equipment, please consult your site Intrinsically Safe approvals. These Half Masks are NOT certified intrinsically safe.