Learn More
Clinofibrate, MedChemExpress
MedChemExpress Clinofibrate (S-8527) is a hypelipidemic agent and a HMG-CoA reductase inhibitor.
Brand:  Medchem Express HY-13528
Encompass_Preferred
 View more versions of this product
Product Code. 16446216
  • 3645.78 DKK / 100mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
Clinofibrate (S-8527) is a hypelipidemic agent and a HMG-CoA reductase inhibitor.
Specifications
Specifications
Clinofibrate
White
C28H36O6
S-8527
CCC(OC1=CC=C(C2(C3=CC=C(OC(CC)(C)C(O)=O)C=C3)CCCCC2)C=C1)(C)C(O)=O
468.58
Research
Metabolism-sugar/lipid metabolism
30299-08-2
Research
100 mg
DMSO : ≥ 30 mg/mL (64.02 mM)
468.58
99.7%
Solid
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.