missing translation for 'onlineSavingsMsg'
Learn More

CLOCK Rabbit anti-Human, Mouse, Rat, Clone: 3D8H6, Novus Biologicals™

Rabbit Monoclonal Antibody

Brand:  Bio-Techne NBP3-16656-100UL

 View more versions of this product

Product Code. 18310605

  • 3692.13 DKK / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

CLOCK Monoclonal antibody specifically detects CLOCK in Human, Mouse, Rat samples. It is validated for Western Blot, Immunofluorescence
Specifications

Specifications

CLOCK
Monoclonal
Unconjugated
BHLHE8, circadian locomoter output cycles protein kaput, Class E basic helix-loop-helix protein 8, clock (mouse) homolog, clock homolog (mouse), EC 2.3.1.48, hCLOCK, KAT13D, KIAA0334bHLHe8circadian locomoter output cycles kaput protein
A synthetic peptide corresponding to a sequence within amino acids 100-200 of human CLOCK (O15516).
100 μg
Cancer, Chromatin Research, Circadian Rhythm, Hypoxia, Neuroendocrinology, Neuroscience, Signal Transduction, Transcription Factors and Regulators, Vision
9575
Store at -20°C. Avoid freeze-thaw cycles.
IgG
Western Blot, Immunofluorescence
3D8H6
PBS, 0.05% BSA, 50% glycerol, pH7.3
Rabbit
Affinity purified
RUO
Primary
Human, Mouse, Rat
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers