missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CNR1 293T Cell Overexpression Lysate (Denatured), Abnova
Cell transient overexpression lysate for CNR1
Brand: Abnova H00001268-T01.100uL
This item is not returnable.
View return policy
Description
This gene encodes one of two cannabinoid receptors. The cannabinoids, principally delta-9-tetrahydrocannabinol and synthetic analogs, are psychoactive ingredients of marijuana.
- Cannabinoid receptor 1 (brain)
- Transfected cell line: 293T
- Plasmid: pCMV-CNR1 full-length
- Host: Human
- Theoretical molecular weight: 52.03kDa
- Storage buffer: 1x sample buffer (50mM Tris-HCl, 2% SDS, 10% glycerol, 300mM 2-mercaptoethanol, 0.01% bromophenol blue)
Western Blotting
Specifications
293T | |
CNR1 293T Cell Transient Overexpression Lysate(Denatured) | |
52.03 | |
pCMV-CNR1 full-length | |
100 μL | |
Human |
Western Blot | |
NM_016083 | |
6.8 | |
Transient overexpression cell Lysate was tested with Anti-CNN2 antibody (H00001265-B01) by western blotting | |
1X Sample Buffer (50 mM Tris-HCl, 2% SDS, 10% glycerol, 300 mM 2-mercaptoethanol, 0.01% Bromophenol blue) | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction