Learn More
Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers.
Brand:  Thermo Scientific™ FLN4567
 View more versions of this product
Product Code. 11306401
  • 19045.59 DKK / Each
Estimated Shipment: 25-07-2025
Description
Ensure optimal security for critical samples by using Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers, for use with Thermo Scientific™ TSX, Forma™, HERAfreeze™, Revco™ and TS-Series freezers.
Features:
  • LN2 backup system maintains temperature down to -80°C with liquid nitrogen
  • CO2 backup system maintains temperature down to -7° or -70°C (model dependant) with CO2
Ordering Information:
Available as field- or factory-installed. Models for UxF300/88300/HFU300T/TSU300 Series are standalone. Field-installed units must be installed by qualified professionals.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
LN2 Backup, Field Installed
-80°C
Revco UxF, HERAfreeze HFU-T, Thermo Scientific TSU Series
Videos
  • Thermo Scientific - Revco UxF freezers
    To the rest of the world, these are samples. To us, they are a scientist's life's work. A lab's mission. A company's breakthrough. We are obsessed with sample integrity.
    Video Player is loading.
    Current Time 0:00
    Duration 0:00
    Loaded: 0%
    Stream Type LIVE
    Remaining Time 0:00
    1x
    • Chapters
    • descriptions off, selected
    • captions off, selected
      Thermo Scientific Revco UxF freezers
    • Thermo Scientific - Revco UxF freezers
      To the rest of the world, these are samples. To us, they are a scientist's life's work. A lab's mission. A company's breakthrough. We are obsessed with sample integrity.
      Video Player is loading.
      Current Time 0:00
      Duration 0:00
      Loaded: 0%
      Stream Type LIVE
      Remaining Time 0:00
      1x
      • Chapters
      • descriptions off, selected
      • captions off, selected
        Thermo Scientific Revco UxF freezers
      Certificates
      Promotions
      Promotions
      Product Content Correction
      Product Title
      Thermo Scientific™ CO2 and LN2 Backup Systems for Ultra-Low Temperature Freezers > LN2 Backup System; Field Installed; For use with Forma 88000, HERAfreeze HFU-T, Revco UxF and TSU Series