Learn More
Buerkle™ Thread adapters LaboPlastBuerkle™ 3/4 in.
StopCock spigots, ball valves and other accessories can be connected to most commercially available containers, with the LaboPlast™ thread adapters.
Brand: Buerkle™ 0600-0120
121.24 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
This item is not returnable.
View return policy
Description
- All adapters have a standard outer thread R 3/4 in.
Specifications
Polypropylene, Polyethylene | |
3/4 in. Inflowing, 2 in. Mauser Outflowing | |
Green/White | |
Thread Adapter |
Product Content Correction
Product Title
Buerkle™ Thread adapters LaboPlastBuerkle™ 3/4 in. > Thread Size: 3/4 in. Inflowing, 2 in. Mauser outflowing
Spot an opportunity for improvement?Share a Content Correction