Learn More
Buerkle™ Thread adapters LaboPlastBuerkle™ 3/4 in.
StopCock spigots, ball valves and other accessories can be connected to most commercially available containers, with the LaboPlast™ thread adapters.
Brand:  Buerkle™ 0600-0020
 View more versions of this product
Product Code. 10558731
  • 54.96 DKK / Each
Only null left
Estimated Shipment: 12-08-2025
Add to basket
This item is not returnable. View return policy
Description
Description
  • All adapters have a standard outer thread R 3/4 in.
Specifications
Specifications
Polypropylene
Kautex External, 3/4 in. Internal
Blue
Thread Adapter
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Buerkle™ Thread adapters LaboPlastBuerkle™ 3/4 in. > Thread Size: 3/4 in. Inflowing, Kautex outflowing