Learn More
COD Standard, 10,000mg/L, Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
500 mL
Specifications
Specifications
Quantity | 500 mL |
Concentration | 10000 mg/L |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
COD10M25C1 | Certificate of Analysis | 26/03/2025 | COD10M |
COD10M24E1 | Certificate of Analysis | 23/07/2024 | COD10M |
COD10M23J1 | Certificate of Analysis | 07/02/2024 | COD10M |
COD10M22B1 | Certificate of Analysis | 20/10/2022 | COD10M |
COD10M21E1 | Certificate of Analysis | 13/01/2022 | COD10M |
Product Content Correction
Product Title
COD Standard, 10,000mg/L, Reagecon™ >
Spot an opportunity for improvement?Share a Content Correction