missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Columbia Agar with Blood / Chocolate Agar

Read more results from a patient sample when you isolate and cultivate fastidious and non-fastidious microorganisms on a bi-plate containing two separate media formulations.

Brand:  Thermo Scientific™ PB5202E

Product Code. 10067162

  • 129.92 DKK / Pack of 10
In Stock
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Isolate and cultivate fastidious and non-fastidious microorganisms from different specimens using Columbia Agar with Blood / Chocolate Agar, a single bi-plate that includes two media formulations.

Read more results from a single patient sample and produce more diagnostic details of pathogenic organisms whilst reducing storage capacity issues in the incubator with a bi-plate that includes two widely used microbiological media.

Columbia Agar with Blood on one side of the plate contains 5% sheep blood to promote growth of fastidious organisms with clearly visible hemolysis forms for staphylococci and streptococci, and it gives typical growth for S.pneumoniae (dent morphology).

Chocolate Agar on the other side of the plate is a highly nutritious medium for the isolation and cultivation of fastidious microorganisms such as Neisseria and Haemophilus spp. The presence of starch ensures that toxic metabolites produced by Neisseria are absorbed, and phosphate buffers are included to prevent changes in pH due to amine production that would affect the survival of the organism.

Not all products are available for sale in all territories. Please inquire.

Remel™ and Oxoid™ products are now part of the Thermo Scientific brand, combining powerful manual, semi-automated and fully automated test products and a comprehensive line of media and diagnostic products to offer a complete, end-to-end solution to quickly deliver the products you need and the quality results your laboratory depends on.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

COL + SB: Carmine red, opaque. CHOC: Signal brown, opaque
Columbia Agar with Blood / Chocolate Agar
Germany
17g ± 5%
10/Pk.
2° to 12°C, in the dark
90mm biplate
Agar
Flow Wrapped - Single Layer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number6262618 Certificate TypeCertificate of Analysis Date28/04/2025 Catalog NumberPB5202E
Lot Number6260892 Certificate TypeCertificate of Analysis Date25/04/2025 Catalog NumberPB5202E
Lot Number6254989 Certificate TypeCertificate of Analysis Date07/04/2025 Catalog NumberPB5202E
Lot Number6252004 Certificate TypeCertificate of Analysis Date31/03/2025 Catalog NumberPB5202E
Lot Number6249109 Certificate TypeCertificate of Analysis Date24/03/2025 Catalog NumberPB5202E
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Columbia Agar with Blood / Chocolate Agar >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.