Learn More
Combined Acid Buffer, Reagecon
BIOPHARMA
Convenient, combined acid buffer solution.
Brand:  Reagecon CAB05
Product Code. 11593052
  • 413.92 DKK / 500mL
Description
  • Combined acid buffer solution
Specifications
Specifications
Liquid
500 mL
HDPE Bottle GL45
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Show More
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberCAB0524K1 Certificate TypeCertificate of Analysis Date28/05/2025 Catalog NumberCAB05
Lot NumberCAB0524C1 Certificate TypeCertificate of Analysis Date12/11/2024 Catalog NumberCAB05
Lot NumberCAB0523C1 Certificate TypeCertificate of Analysis Date31/03/2024 Catalog NumberCAB05
Lot NumberCAB0522F1 Certificate TypeCertificate of Analysis Date04/10/2023 Catalog NumberCAB05
Lot NumberCAB0521J1 Certificate TypeCertificate of Analysis Date30/04/2022 Catalog NumberCAB05
Promotions
Promotions
Product Content Correction
Product Title
Combined Acid Buffer, Reagecon > 500 mL