Learn More
Novus Biologicals™ Complement Component C2 Antibody (ARGX-117) - Humanized, Novus Biologicals™
Human Monoclonal Antibody
Brand:  Novus Biologicals™ NBP3-28215-50ug
 View more versions of this product
Product Code. 30045951
  • 1525.00 DKK / 50µg
This item is not returnable. View return policy
Description
Description
Complement Component C2 Monoclonal antibody specifically detects Complement Component C2 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Complement Component C2
Monoclonal
Unconjugated
P06681
Human
Protein A purified
RUO
717
Human
Purified
Flow Cytometry, ELISA, Functional Assay
ARGX-117
Lyophilized from 25mM histidine, 8% sucrose, 0.01% Tween80 (pH6.2)
C3/C5 convertase, CO2, complement C2, complement component 2, complement component C2, DKFZp779M0311, EC 3.4.21, EC 3.4.21.43
Complement C2
50 ÎĽg
Primary
Reconstitute with sterile, distilled water to a final concentration of 1 mg/mL. Gently shake to solubilize completely. Do not vortex.
Store at -20°C in powder form. Store at -80°C once reconstituted.
IgG1
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Novus Biologicals™ Complement Component C2 Antibody (ARGX-117) - Humanized, Novus Biologicals™ > 50 ÎĽg, Unconjugated