Learn More
SI Analytics™ Complete Dosing Unit
SI Analytics™ Complete Dosing Unit comes with piston and piston rod, as well as a UV protection coat for use with the TITRONIC™ Series.
2510.00 DKK - 2865.00 DKK
Specifications
Type | Complete Dosing Unit |
---|---|
Includes | Piston, Piston Rod and UV Protection Coat |
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Volume (Metric) | Price | Quantity & Availability | ||||||
10267871
|
SI Analytics™
285223718 |
20 mL |
2510.00 DKK
Each |
In Stock
Log in to see stock. |
||||||
10499821
|
SI Analytics™
285223767 |
50 mL |
2865.00 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
- Dosing unit
- Includes piston, piston rod, and UV protection coat
- For use wtih TITRONIC™ Universal and TITRONIC™ 300
Specifications
Complete Dosing Unit | |
Piston, Piston Rod and UV Protection Coat |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
SI Analytics™ Complete Dosing Unit