Learn More
Lenz Laborglasinstrumente™ Condenser
Condenser for condensing vapors.
Brand:  Lenz Laborglasinstrumente™ 05210414
 View more versions of this product
Product Code. 10060142
  • 377.00 DKK / Each
Only null left
Estimated Shipment: 22-08-2025
Description
  • Chemical resistance
  • High temperature resistance
Specifications
Specifications
400 mm
1 Ea.
NS 29/32
Condenser
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Condenser > Neck Size: 29/32; Length: 400 mm