Learn More
Conductivity Standards 10μS/cm, Mettler Toledo™
Brand:  Mettler Toledo 51300169
805.78 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Quantity 250 mL
Product Code. 10545284
  • 1245.00 DKK / 250mL
Specifications
10ÎĽS/cm
250 mL
Documents
One moment while we fetch your results.
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Conductivity Standards 10μS/cm, Mettler Toledo™ > Quantity: 250mL