Learn More
Lauda™ Connecting Cable
Material: Stainless Steel; For Use With: 2 Lemo plugs
Brand:  Lauda™ UK 246
Product Code. 10008011
  • 1860.00 DKK / Each
Explore more special offers
Description
Description
  • For use with external control on all units with Lemo plug
Specifications
Specifications
2 Lemo plugs
Connecting Cable
Stainless Steel
2.5 mm
Documents
One moment while we fetch your results.
Product Title
Lauda™ Connecting Cable > LengthMetric:2.5mm