Learn More
SCAT Connectors for HPLC
Safe connection of tubes of different sizes: directly and without technical aids. For safe collection of liquid HPLC waste, and many other applications in the laboratory!
Brand:  SCAT 107816
76.72 DKK valid until 2025-08-24
Use promo code "24132" to get your promotional price.
 View more versions of this product
Product Code. 10024257
  • 94.75 DKK / Each
Everyday Lab Essentials
Use promo code 24132 at checkout
View Available Promotions
Only null left
Estimated Shipment: 30-07-2025
Description
SCAT™ connectors are available in the following versions:
  • Conical connectors
  • Y-connectors
  • Tube connectors
  • Capillary connectors
Specifications
Specifications
6.2 to 7.5 mm
Tubing
Tube Connector
Polypropylene
HPLC/UHPLC Systems
Certificates
Promotions
Promotions
  • Multiple Manufacturers
    From wipes to waste bins, labels to liners – save on them all.
    Promo Code: 24132
    Expires2025-08-24
    New Offer
    Shop Now
Product Content Correction
Product Title
SCAT Connectors for HPLC > Connectors for HPLC; Type: Tube Connector; Material: Polypropylene; Diameter (Metric): 6.2 to 7.5 mm; For Use With: Tubing