Learn More
SymLine Continuous Level Control Accessories
Accessories for Continuous Level Control
Brand: SymLine 108178
2045.03 DKK valid until 2025-08-24
Use promo code "24132" to get your promotional price.
This item is not returnable.
View return policy
Description
Measuring filling levels continuously - Monitoring non-transparent canisters on a permanent basis - No problem with the continuous level control unit!Specifications
For proper functionality use in combination with isolating switching amplifier 108278 & Canister 108042-S1. Housing M18 x 1, active surface material PTFE, housing material brass. Flush mountable. Sensing distance 1 to 8 mm adjustable. | |
ATEX, II 2 G EEx ia IIC T1-T6, NAMUR | |
Capacitive Rod Sensor |
Product Content Correction
Product Title
SymLine Continuous Level Control Accessories > Type: Continuous level control; Capacitive rod sensor
Spot an opportunity for improvement?Share a Content Correction