missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brady™ Continuous Vinyl Tape for BradyJet J4000 and J5000
B-2595 is ideal for indoor facility labels, including 5S, safety, warning, pipe markers, equipment and visual workplace labels on stainless steel and smooth surfaces.
2220.00 DKK - 4115.00 DKK
Specifications
Autoclavable | Not Autoclavable |
---|---|
Adhesive Type | Permanent Acrylic |
Color | White |
Finish | Matte |
Material | Vinyl (Inkjet-receptive) |
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Width (Metric) | Price | Quantity & Availability | |||||
17489182
|
Brady™
148656 |
57 mm |
2220.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
17459182
|
Brady™
148653 |
101 mm |
3060.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
17489192
|
Brady™
148666 |
152 mm |
3680.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
17459192
|
Brady™
148663 |
203.2 mm |
4115.00 DKK
Each |
In Stock
Log in to see stock. |
|||||
Description
- Specially-formulated topcoat enhances colors and label appearance
- Tested to withstand exposure to liquids, alcohol and chemicals, as well as indoor fluorescent and UV lighting
- Topcoat protects print from smudging and harsh conditions
- Inkjet-receptive labels for use with the BradyJet J4000 or J5000 Color Label Printer
- An ink cartridge is required to print this material
Specifications
Not Autoclavable | |
White | |
Vinyl (Inkjet-receptive) | |
Non-sterile | |
BradyJet J4000 and J5000 Color Label Printers | |
1 Tape | |
0.14 mm |
Permanent Acrylic | |
Matte | |
Ink Jet | |
Electrical Labeling, Hazard Communication Identification, Warehouse Marking | |
1 | |
-40°C to +60°C | |
30 m |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title