missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Serva Electrophoresis™ Coomassie™ Brilliant Blue R 250
324.64 DKK - 802.61 DKK
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11832020
|
Serva Electrophoresis™
1752501 |
25 g |
324.64 DKK
Each |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||||
11842020
|
Serva Electrophoresis™
1752502 |
100 g |
802.61 DKK
Each |
Estimated Shipment: 08-10-2024
Log in to see stock. |
|||||
Specifications
Dark Blue | |
Protein Stain |