Learn More
Copper Phosphorus shot, Thermo Scientific Chemicals
Código de producto. 11369249
missing translation for 'orderingAttributeHoverText'
Quantity:
500 g
2 kg
Este artículo no se puede devolver. Vea la política de devoluciones
Este artículo no se puede devolver. Vea la política de devoluciones
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Identificadores de productos químicos
MDL Number MFCD01074956
Especificaciones
Quantity 500 g
Chemical Name or Material Copper Phosphorus shot