Learn More
Copper Phosphorus shot, Thermo Scientific Chemicals
Product Code. 11379249
Click to view available options
Quantity:
500 g
2 kg
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
MDL Number MFCD01074956
Specifications
Quantity 2 kg
Chemical Name or Material Copper Phosphorus shot