Learn More
Copper powder, spherical, -100 mesh, 99.5% (metals basis) Product Code.: 11336078
Copper powder, spherical, -100 mesh, 99.5% (metals basis)
Product Code. 11336078
1 kg
Click to view available options
Quantity:
50 g
250 g
1 kg
5 x 1 kg
Product Code. 11336078
Brand: Thermo Scientific Alfa Aesar 011070.A1
Request Bulk or Custom Format
CAS: 7440-50-8 | Cu | 63.55 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]
Specifications
UN Number UN3077
Quantity 1 kg
Percent Purity 99.5%
Mesh Size −100 Mesh
Odor Odorless
Form Spherical Powder
Assay (metals basis)
Chemical Name or Material Copper powder
RUO – Research Use Only
Product Content Correction
Product Title
Copper powder, spherical, -100 mesh, 99.5% (metals basis) >