missing translation for 'onlineSavingsMsg'
Learn More
Copper wire, 2.0mm (0.08in) dia, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals Product Code.: 11301568

Copper wire, 2.0mm (0.08in) dia, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals

Product Code. 11301568
150 cm
Click to view available options
Quantity:
50 cm
150 cm
500 cm
This item is not returnable. View return policy

Product Code. 11301568

Brand: Thermo Scientific Alfa Aesar 010975.BZ

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Form: Approx. 28.1g/m

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-50-8
Molecular Formula Cu
Molecular Weight (g/mol) 63.55
MDL Number MFCD00010965
InChI Key RYGMFSIKBFXOCR-UHFFFAOYSA-N
Synonym cuprum, cobre, cuivre, blister, cathode, bronze, powder, anode, precipitates, kupfer
PubChem CID 23978
ChEBI CHEBI:30052
IUPAC Name copper
SMILES [Cu]

Specifications

Appearance Copper colored wire
Density 8.94g/cm3
Assay Percent Range 99.999% (Metals basis)
Quantity 150 cm
Odor Odorless
Form Wire
Chemical Name or Material Copper

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY05K003 Certificate TypeCertificate of Analysis Date17/12/2024 Catalog Number010975.BZ,010975.BW,010975.CC
Lot NumberN14K006 Certificate TypeCertificate of Analysis Date22/02/2024 Catalog Number010975.BZ,010975.BW,010975.CC
Lot NumberX28I007 Certificate TypeCertificate of Analysis Date15/11/2022 Catalog Number010975.BZ,010975.BW,010975.CC
Lot NumberQ26I063 Certificate TypeCertificate of Analysis Date11/08/2022 Catalog Number010975.BZ,010975.BW,010975.CC
Lot NumberT22I034 Certificate TypeCertificate of Analysis Date08/08/2022 Catalog Number010975.BZ,010975.BW,010975.CC

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Copper wire, 2.0mm (0.08in) dia, Puratronic™, 99.999% (metals basis), Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.