Learn More
Corning™ CellBIND™ 1536-Well, CellBIND-Treated, Flat-Bottom Microplate
Color:White; Sterility:Non-sterile; Lid:Without Lid
Brand:  Corning™ 4573
Product Code. 17730708
  • 12781.63 DKK / Pack of 50
Estimated Shipment: 03-01-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Corning 1536-well microplates are our highest density microplates available for high throughput screening. The microplates conform to standard microplate footprint and dimensions. These microplates are offered in solid black and white polystyrene, with round or flat bottoms, and in black-clear bottom formats.
  • 127 μm film thickness
  • 536-well low base, black/clear or white/clear bottom microplates
  • Bar coded
  • Custom bar codes available for compatibility with the Kalypsys system and with UHTS systems
  • Low auto-fluorescence
  • Broad chemical resistance including DMSO and alcohol
  • High mechanical stability
  • Optimized for flatness and uniformity
  • Low birefringence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Includes Barcode
Without Lid
32 x 48
White
Cycloolefin Copolymer
8 μL
CellBIND
For higher sensitivity
10/Pk.
Standard
0.03 cm2
Corning CellBIND Surface
Automated dispensers
1536
Non-sterile
Flat Bottom (F-well)
50/Cs.
50/Cs.