Learn More
Corning™ 384-Well Black, Clear Bottom Ultra-Low Attachment Microplate
Use for both top and bottom reading instruments
Brand:  Corning™ 3827
Product Code. 13237639
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
This item is not returnable. View return policy
Description
Description
  • Covalently bound hydrogel layer that effectively inhibits cellular attachment
  • Surface minimizes protein absorption, enzyme activation and cellular activation
  • Surface is noncytotoxic, biologically inert and nondegradable
  • Sterilized by gamma irradiation and certified nonpyrogenic
  • Black wall microplates have low background fluorescence, minimal light scatter and reduced crosstalk
  • Opaque walls to prevent well-to-well crosstalk
  • Optically clear flat well bottom permits direct microscopic viewing
  • Bulk packed 20 per bag
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Standard
0.16 cm2
Ultra-Low Attachment (covalently bound Hydrogel layer)
Both top and bottom reading instruments
384
Sterile
Effectively inhibits cellular attachment, minimizes protein absorption, enzyme activation and cellular activation
With Lid
16 x 24
Black
Polystyrene
20 to 80 μL
Flat Bottom (F-well)
Clear
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
One moment while we fetch your results.
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ 384-Well Black, Clear Bottom Ultra-Low Attachment Microplate > 112μL; Black; Sterile