Learn More
Corning™ 384-Well, Cell Culture-Treated, Flat-Bottom Microplate
Color:Black; Sterility:Sterile; Lid:With Lid
Brand: Corning™ 3822
This item is not returnable.
View return policy
Description
Designed to reduce well-to-well cross-talk during fluorescent and luminescent assays.
Specifications
No Barcode | |
With Lid | |
16 x 24 | |
Polystyrene | |
5 to 40 μL | |
Cell Culture | |
Provides easier access and imaging for superior assay performance, fluorescence applications, reduce cross-contamination during assay and liquid-handling process | |
10/Pk. |
Standard | |
0.03 cm2 | |
Black | |
384 | |
Sterile | |
Flat Bottom (F-well) | |
50/Cs. | |
Black |
COR001 Lifescience Guid
File size: 7 MB
Product Content Correction
Product Title
Corning™ 384-Well, Cell Culture-Treated, Flat-Bottom Microplate > Low Volume, with Lid; Black; Flat; TC-treated; Yes
Spot an opportunity for improvement?Share a Content Correction