Learn More
Corning™ 384-Well, Cell Culture-Treated, Flat-Bottom Microplate
GREENER_CHOICE
Color:Black; Sterility:Sterile; Lid:With Lid
Brand:  Corning™ 3822
Product Code. 10067272
  • 4483.35 DKK / Pack of 50
Estimated Shipment: 27-08-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
Designed to reduce well-to-well cross-talk during fluorescent and luminescent assays.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
No Barcode
With Lid
16 x 24
Polystyrene
5 to 40 μL
Cell Culture
Provides easier access and imaging for superior assay performance, fluorescence applications, reduce cross-contamination during assay and liquid-handling process
10/Pk.
Standard
0.03 cm2
Black
384
Sterile
Flat Bottom (F-well)
50/Cs.
Black
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
Special Offers
Special Offers
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Corning™ 384-Well, Cell Culture-Treated, Flat-Bottom Microplate > Low Volume, with Lid; Black; Flat; TC-treated; Yes