Learn More
Corning™ Collection Bags
Collection bags with three ports used for biopharmaceutical applications
408.57 DKK - 2604.68 DKK
Specifications
Tubing Style | Heat sealable flexible tubing |
---|---|
No. per Case | 1/Ea. |
No. per Pack | 1/Pk. |
Quantity | Each |
Product Code | Brand | Capacity (Metric) | Film Description | Port Detail | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Film Description | Port Detail | Price | Quantity & Availability | |||
17283574
|
Corning™
91-200-42 |
1 L | EVA | Male MPC connector with 0.25 in. barb, female MPC cap, female MPC connector with 0.25 in. barb, male MPC plug, break-off injection site |
424.05 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17273574
|
Corning™
91-200-41 |
0.5 L | EVA | Male MPC connector with 0.25 in. barb, female MPC cap, female MPC connector with 0.25 in. barb, male MPC plug, break-off injection site |
408.57 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17213584
|
Corning™
91-200-82 |
100 L | ULDPE | 60 L x 0.5 I.D. x 0.75 in. O.D. and 40 L x 0.12 I.D. x 0.25 in. O.D. chemically resistant, heat sealable flexible tubings, female luer with 0.15 in. barb, swabbable valve, non-breather female luer cap with female MPX cap and male MPX connector with 0.5 in |
2477.17 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
17223584
|
Corning™
91-200-83 |
200 L | ULDPE | 60 L x 0.5 I.D. x 0.75 in. O.D. and 40 L x 0.12 I.D. x 0.25 in. O.D. chemically resistant, heat sealable flexible tubings, female luer with 0.15 in. barb, swabbable valve, non-breather female luer cap with female MPX cap and male MPX connector with 0.5 in |
2604.68 DKK
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
Corning collection bags are available in 2D hanging configurations with multiple bag volumes and connector configurations.
Spezifikation
Heat sealable flexible tubing | |
1/Ea. | |
1/Pk. | |
Each |
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfĂĽllen.
Name des Produkts
Corning™ Collection Bags