Learn More
Corning™ RPMI 1640 Medium (Mod.) 1X with L-Glutamine Product Code.: 15303541
Corning™ RPMI 1640 Medium (Mod.) 1X with L-Glutamine
BIOPHARMA
Product Code. 15303541
6 × 500 mL, Glutamine, None, Pack of 6
Click to view available options
Quantity:
6 × 500 mL
With Additives:
Glutamine
Without Additives:
None
Unit Size:
Pack of 6
Product Code. 15303541
Brand: Corning™ 10040CV
Guaranteed low endotoxin levels ≤0.025EU/mL
  • Ready-to-use media for mammalian in vitro cell culture is supplied in polyethylene teraphthalate (PET) plastic bottles
  • Sonically welded cap liners eliminate toxic resins or glues that could leach into media
  • Shipped at ambient temperature
TRUSTED_SUSTAINABILITY
Specifications
Form Liquid
Product Type RPMI 1640
With Additives Glutamine
Without Additives None
Quantity 6 × 500 mL
pH 7.2 to 7.4
Content And Storage 2° to 8°C
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number29924004 Certificate TypeCertificate of Quality Date18/07/2025 Product Code10-040-CV
Lot Number31724011 Certificate TypeCertificate of Quality Date06/03/2025 Product Code10-040-CV
Lot Number17724019 Certificate TypeCertificate of Quality Date27/02/2025 Product Code10-040-CV
Lot Number03724008 Certificate TypeCertificate of Quality Date29/01/2025 Product Code10-040-CV
Lot Number03724008 Certificate TypeCertificate of Analysis Date17/07/2024 Product Code10-040-CV
Product Content Correction
Product Title
Corning™ RPMI 1640 Medium (Mod.) 1X with L-Glutamine >