Learn More
Reagent Grade Water Tested to USP Purified Water, Corning™ Product Code.: 15313711
Reagent Grade Water Tested to USP Purified Water, Corning™
BIOPHARMA
Product Code. 15313711
2 x 4 L
Click to view available options
Quantity:
2 x 4 L
Product Code. 15313711
Brand: Corning Media Tech 46002LF
Used in common laboratory applications requiring highly pure water. Corning™ Reagent Grade Water Tested to USP Purified Water is used in the preparation of reagents, rinsing glassware and plastic ware.
  • Tested to USP monograph for purified water
  • DNase, RNase, Protease-free
  • Endotoxin levels ≤0.025EU/mL
Certifications
Manufactured under cGMP (21 CFR 820) in ISO 13485 certified facility
TRUSTED_SUSTAINABILITY
Specifications
Quantity 2 x 4 L
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number24024006 Certificate TypeCertificate of Quality Date22/04/2025 Catalog Number46-002-LF
Lot Number01624005 Certificate TypeCertificate of Quality Date24/07/2024 Catalog Number46-002-LF
Product Content Correction
Product Title
Reagent Grade Water Tested to USP Purified Water, Corning™ >