Learn More
Corning™ Collagen I, Bovine Product Code.: 11533550
Corning™ Collagen I, Bovine
BIOPHARMAGREENER_CHOICE
Product Code. 11533550 Shop All Corning Products
30 mg, 30mg
Click to view available options
Quantity:
30 mg
Unit Size:
30mg
Product Code. 11533550
Brand: Corning™ 354231
Used as either a thin layer on tissue-culture surfaces to enhance cell attachment and proliferation, or as a gel to promote expression of cell-specific morphology and function
Corning extracellular matrices (ECMs) enable researchers to mimic in vivo environments for 2D and 3D cell culture applications.
Corning offers a broad range of collagen types that are derived from multiple species. These collagens have been used to culture a variety of cell types to promote adhesion, growth, and/or differentiation.
Greener Choice Claims
  • Environmental benefits include:
    • Renewable Energy
    • Certifications and Seals of Approval
TRUSTED_SUSTAINABILITY
Specifications
Concentration 5 to 10 ÎĽg/cm2
For Use With (Application) Cell Attachment and Proliferation, Cell-specific Morphology and Function
Formulation Liquid in 0.012N hydrogen chloride
Quantity 30 mg
Source Bovine Dermis
Product Type Collagen, Bovine Type I
Content And Storage 2°C to 8°C
Purity or Quality Grade >95% by SDS-PAGE
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Product Code
Lot Number28324003 Certificate TypeCertificate of Quality Date15/07/2025 Product Code354231
Lot Number06625001 Certificate TypeCertificate of Quality Date15/07/2025 Product Code354231
Lot Number24823007 Certificate TypeCertificate of Quality Date05/12/2024 Product Code354231
Lot Number09624001 Certificate TypeCertificate of Quality Date26/11/2024 Product Code354231
Lot Number2325001 Certificate TypeCertificate of Quality Date17/08/2023 Product Code354231
Product Content Correction
Product Title
Corning™ Collagen I, Bovine >